Merck
  • Constitutive and LPS-stimulated secretome of porcine Vascular Wall-Mesenchymal Stem Cells exerts effects on in vitro endothelial angiogenesis.
Constitutive and LPS-stimulated secretome of porcine Vascular Wall-Mesenchymal Stem Cells exerts effects on in vitro endothelial angiogenesis.
BMC veterinary research (2019-04-29)
Chiara Bernardini, Martina Bertocchi, Augusta Zannoni, Roberta Salaroli, Irvin Tubon, Giovanni Dothel, Mercedes Fernandez, Maria Laura Bacci, Laura Calzà, Monica Forni
ABSTRACT
MSCs secretome is under investigation as an alternative to whole-cell-based therapies, since it is enriched of bioactive molecules: growth factors, cytokines and chemokines. Taking into account the translational value of the pig model, the leading aim of the present paper was to characterize the secretome of porcine Vascular Wall-Mesenchymal Stem Cells (pVW-MSCs) and its change in presence of LPS stimulation. Moreover, considering the importance of angiogenesis in regenerative mechanisms, we analysed the effect of pVW-MSCs secretome on in vitro angiogenesis. Our results demonstrated that conditioned medium from unstimulated pVW-MSCs contained high levels of IL-8, GM-CSF, IFN-γ and other immunomodulatory proteins: IL-6 IL-18 IL-4 IL-2 IL-10. LPS modulates pVW-MSCs gene expression and secretome composition, in particular a significant increase of IL-6 and IL-8 was observed; conversely, the amount of GM-CSF, IFN-γ, IL-2, IL-4, IL-10 and IL-18 showed a significant transient decrease with the LPS stimulation. Conditioned medium from unstimulated pVW-MSCs induced in vitro endothelial angiogenesis, which is more evident when the conditioned medium was from LPS stimulated pVW-MSCs. The lines of evidence here presented shed a light on possible future application of secretome derived by pVW-MSCs on research studies in translational regenerative medicine.