Merck
  • USP14 inhibits ER-associated degradation via interaction with IRE1alpha.
USP14 inhibits ER-associated degradation via interaction with IRE1alpha.
Biochemical and biophysical research communications (2009-01-13)
Atsushi Nagai, Hisae Kadowaki, Takeshi Maruyama, Kohsuke Takeda, Hideki Nishitoh, Hidenori Ichijo
ABSTRACT
Accumulation of unfolded proteins within the endoplasmic reticulum (ER) lumen induces ER stress. Eukaryotic cells possess the ER quality control systems, the unfolded protein response (UPR), to adapt to ER stress. IRE1alpha is one of the ER stress receptors and mediates the UPR. Here, we identified ubiquitin specific protease (USP) 14 as a binding partner of IRE1alpha. USP14 interacted with the cytoplasmic region of IRE1alpha, and the endogenous interaction between USP14 and IRE1alpha was inhibited by ER stress. Overexpression of USP14 inhibited the ER-associated degradation (ERAD) pathway, and USP14 depletion by small interfering RNA effectively activated ERAD. These findings suggest that USP14 is a novel player in the UPR by serving as a physiological inhibitor of ERAD under the non-stressed condition.