Merck
  • Early Fesoterodine Fumarate Administration Prevents Neurogenic Detrusor Overactivity in a Spinal Cord Transected Rat Model.
Early Fesoterodine Fumarate Administration Prevents Neurogenic Detrusor Overactivity in a Spinal Cord Transected Rat Model.
PloS one (2017-01-07)
Xavier Biardeau, Mikolaj Przydacz, Shachar Aharony, George Loutochin, Lysanne Campeau, Maeva Kyheng, Jacques Corcos
ABSTRACT
In spinal cord injury, onset of detrusor overactivity (DO) is detrimental for quality of life (incontinence) and renal risk. Prevention has only been achieved with complex sophisticated electrical neuromodulation techniques. To assess the efficacy of early fesoterodine fumarate (FF) administration in preventing bladder overactivity in a spinal cord transected (SCT) rat model. 33 Sprague-Dawley rats were allocated to 6 groups-Group 1: 3 normal controls; Group 2: 6 SCT controls; Group 3: 6 SCT rats + FF 0.18 mg/kg/d; Group 4: 6 SCT rats + FF 0.12 mg/kg/d; Group 5: 6 SCT rats + FF 0.18 mg/kg/d + 72-h wash-out period; Group 6: 6 SCT rats + FF 0.12 mg/kg/d + 72-h wash-out period. SCT was performed at T10. FF was continuously administered. Cystometry was undertaken 6 weeks after SCT in awake rats recording intermicturition pressure (IMP), baseline pressure, threshold pressure (Pthres) and maximum pressure (Pmax). Normal controls and SCT controls were initially compared using the Mann-Whitney U tests in order to confirm the SCT effect on cystometric parameters. The comparisons in cystometric and metabolic cage parameters between SCT controls and treated rats were done using post-hoc Dunn's tests for Kruskal-Wallis analysis. Statistical testing was conducted at the two-tailed α-level of 0.05. Pressure parameters were significantly higher in SCT control group compared to normal controls. Six weeks after SCT, IMP was significantly lower in low dose treated group than in SCT controls. Pmax was significantly lower in 3 treated groups compared to SCT controls. Pthres was significantly lower in full time treated groups than in SCT controls. Early administration of FF modulates bladder overactivity in a SCT rat model. Whereas short-term prevention has been demonstrated, the long-term should be further analyzed. Clinical application of these results should confirm this finding through randomized research protocols.
Sigma-Aldrich
Fesoterodine fumarate, ≥93% (HPLC)