Merck
  • (D-Ala2)-Met-enkephalinamide: a potent, long-lasting synthetic pentapeptide analgesic.
(D-Ala2)-Met-enkephalinamide: a potent, long-lasting synthetic pentapeptide analgesic.
Science (New York, N.Y.) (1976-10-15)
C B Pert, A Pert, J K Chang, B T Fong
PMID968485
ABSTRACT
[D-Ala2]-Met-enkephalinamide (DALA), a synthetic enkephalin analog designed by in vitro analysis, binds to opiate receptors almost as tightly as methionine-enkephalin. Since it is not susceptible to degradation by brain enzymes, low doses (5 to 10 micrograms) cause profound, long-lasting, morphine-like analgesia when microinjected into rat brain.