Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of cholesterol depletion and temperature on the isolation of detergent-resistant membranes from human erythrocytes.
Effect of cholesterol depletion and temperature on the isolation of detergent-resistant membranes from human erythrocytes.
The Journal of membrane biology (2010-03-27)
Cleyton C Domingues, Annarita Ciana, Armando Buttafava, Bruna Renata Casadei, Cesare Balduini, Eneida de Paula, Giampaolo Minetti
ABSTRACT
Transient lateral microdomains or lipid rafts play important roles in many physiological membrane-mediated cell processes. Detergent-resistant membranes (DRMs) are good models for the study of lipid rafts. Here we report that DRMs can be obtained by treating human erythrocytes with the nonionic detergents Triton X-100 or octaethylene glycol monododecyl ether (C(12)E(8)) at 37 degrees C, and by treatment at 4 degrees C of cholesterol-depleted erythrocytes. Electron paramagnetic resonance with spin labels inserted at different membrane depths (5- and 16-doxyl stearic acids, 5-SASL and 16-SASL) were used to measure the order parameter (S) of the cell membranes and DRMs. We previously reported significantly higher S values in DRMs with respect to intact erythrocyte membranes. Here we show that higher S values were still measurable in DRMs prepared from intact erythrocytes at 37 degrees C, or from cholesterol-depleted cells at 4 degrees C, for both detergents. For 5-SASL only, increased S values were measured in 4 degrees C DRMs obtained from cholesterol-depleted versus intact erythrocytes. Flotillin-2, a protein marker of lipid rafts, was found in DRMs from intact cells in trace amounts but it was sensitively increased in C(12)E(8) DRMs prepared at 4 degrees C from cholesterol-depleted erythrocytes, while the membrane-skeletal proteins spectrin and actin were excluded from both Triton X-100 and C(12)E(8) DRMs. However, contrary to the 4 degrees C treatment results, flotillin-2 and stomatin were not resistant to Triton X-100 and C(12)E(8) treatment at physiological temperature. The role of cholesterol in DRMs formation is discussed and the results presented provide further support for the use of C(12)E(8) to the study of DRMs.
Sigma-Aldrich
Octaethylene glycol monododecyl ether, ≥98% (GC)