Merck
  • Effect of over-expression of pigment epithelium derived factor (PEDF) on developing retinal vasculature in the mouse.
Effect of over-expression of pigment epithelium derived factor (PEDF) on developing retinal vasculature in the mouse.
Molecular vision (2004-11-18)
Wai T Wong, Tonia S Rex, Alberto Auricchio, Albert M Maguire, Daniel Chung, Waixing Tang, Jean Bennett
ABSTRACT
Pigment epithelium derived factor (PEDF) is a secreted protein with demonstrated anti-angiogenic properties, and with potential application for the treatment of neovascular disease. Delivery of pigment epithelium derived factor to the retina via virus mediated gene transfer has been shown to inhibit neovascularization in a number of experimental models. While pigment epithelium derived factor is endogenously expressed in the retina, its role in guiding normal vessel development and growth is not yet known. This study aimed to determine whether over-expression of pigment epithelium derived factor alters the normal pattern of retinal vessel development. Neonatal (age postnatal day 2 (P2)) CD1 mice were injected subretinally unilaterally with AAV2/1.CMV.PEDF while contralateral eyes were injected subretinally with AAV2/1.CMV.EGFP as control. Cohorts of animals were sacrificed at P7 to P21 and the retinal vasculature was co-labeled through fluorescein-dextran perfusion and immunohistochemistry. Vascular size, localization, and structure were analyzed using light and confocal microscopy. Additional cohorts were use to obtain quantitative levels of pigment epithelium derived factor protein through ELISA. The extent of vessel growth from the optic disk to periphery over time (i.e., the radius of retinal vasculature), and the area of expansion of the neural retina were unaffected by over-expression of pigment epithelium derived factor to levels at least 3.5 fold higher than endogenous levels. The thicknesses of the various retinal layers were similar in AAV2/1.CMV.PEDF treated and control injected eyes. Three dimensional analysis of confocal images shows a slight delay in the rate of growth of vasculature into the deeper layers of the retina in pigment epithelium derived factor treated eyes compared to EGFP treated control eyes. However, the normal differentiation of vessels into arterioles, and venules, and the formation of a capillary network continued to occur, achieving normal and complete maturation of vascular structure by P21. Over-expression of pigment epithelium derived factor in the developing retina exerted no marked or permanent effects on retinal vessel growth and differentiation. The findings are relevant to the safety of the potential therapeutic use of pigment epithelium derived factor in human retinal disease.
Sigma-Aldrich
ChemiKine Pigment Epithelium Derived Factor, Sandwich ELISA, This ChemiKine Pigment Epithelium Derived Factor, Sandwich ELISA is used to measure & quantify PEDF levels in Neuroscience research.