Merck
  • LncRNA snaR upregulates GRB2-associated binding protein 2 and promotes proliferation of ovarian carcinoma cells.
LncRNA snaR upregulates GRB2-associated binding protein 2 and promotes proliferation of ovarian carcinoma cells.
Biochemical and biophysical research communications (2018-08-11)
Yiping Huang, Yingying Hu, Zujian Jin, Zhaojun Shen
ABSTRACT
The functionality of lncRNA snaR has only been characterized in breast cancer and colon cancer. The aim of the current study is to explore the involvement of lncRNA snaR in ovarian carcinoma (OC). Expression of lncRNA snaR and GRB2-associated binding protein 2 (GAB2) in plasma of both patients with OC and healthy females was detected by qRT-PCR. Application value of plasma lncRNA snaR in the diagnosis of OC was evaluated by ROC analysis. Correlation between plasma lncRNA snaR and GAB2 was analyzed by Pearson correlation coefficient. LncRNA snaR and GAB2 expression vectors as well as GAB2 siRNA were transfected into cells of human OC cell lines, and the effect on lncRNA snaR expression, GAB2 expression and cell proliferation was detected by qRT-PCR, western blot and CCK-8 assay. It was observed that plasma levels of lncRNA snaR and GAB2 were significantly higher in OC patients than those in healthy controls. In effect, high levels of plasma lncRNA snaR and GAB2 distinguished OC patients from healthy controls. Plasma lncRNA snaR and GAB2 were positively correlated in OC patients but not in healthy controls. LncRNA snaR overexpression promoted cancer cell proliferation and upregulated GAB2 expression. GAB2 overexpression also promoted cancer cell proliferation but showed no significant effects on lncRNA snaR expression, while GAB2 siRNA silencing significantly attenuated the enhancing effects of lncRNA snaR overexpression on cancer cell proliferation. LncRNA snaR may promote proliferation of ovarian carcinoma cells by upregulating GAB2 expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human GAB2