Merck
  • Soy Phosphatidylglycerol Reduces Inflammation in a Contact Irritant Ear Edema Mouse Model In Vivo.
Soy Phosphatidylglycerol Reduces Inflammation in a Contact Irritant Ear Edema Mouse Model In Vivo.
The Journal of pharmacology and experimental therapeutics (2018-04-27)
Ding Xie, Vivek Choudhary, Mutsa Seremwe, John G Edwards, Angela Wang, Aaron C Emmons, Katherine A Bollag, Maribeth H Johnson, Wendy B Bollag
ABSTRACT
We have previously shown that phosphatidylglycerol (PG) regulates the function of keratinocytes, the predominant cells that compose the epidermis, inhibiting the proliferation of rapidly dividing keratinocytes. In particular, soy PG, a PG mixture with a high proportion of polyunsaturated fatty acids, is efficacious at inhibiting these proliferating keratinocytes. Psoriasis is a skin disorder characterized by hyperproliferation of keratinocytes and inflammation. Data in the lung suggest that PG in pulmonary surfactant inhibits inflammation. To investigate the possibility of using PG containing polyunsaturated fatty acids for the treatment of psoriasis, we examined the effect of soy PG on inflammation induced by the application of 12-O-tetradecanoylphorbol 13-acetate (TPA), a contact irritant, to mouse ears in vivo. We monitored ear thickness and weight as a measure of ear edema, as well as CD45-positive immune cell infiltration. Our results indicate that soy PG when applied together with 1,25-dihydroxyvitamin D3 (vitamin D), an agent known to acutely disrupt the skin barrier, suppressed ear edema and inhibited the infiltration of CD45-positive immune cells. On the other hand, neither PG nor vitamin D alone was effective. The combination also decreased tumor necrosis factor-α (TNFα) levels. This result suggested the possibility that PG was not permeating the skin barrier efficiently. Therefore, in a further study we applied PG in a penetration-enhancing vehicle and found that it inhibited inflammation induced by the phorbol ester and decreased CD45-positive immune cell infiltration. Our results suggest the possibility of using soy PG as a topical treatment option for psoriasis.
Product Description
Avanti
Soy PG, Avanti Research - A Croda Brand 841148P, powder