Merck
  • Chitosan Nanoparticles of Gamma-Oryzanol: Formulation, Optimization, and In vivo Evaluation of Anti-hyperlipidemic Activity.
Chitosan Nanoparticles of Gamma-Oryzanol: Formulation, Optimization, and In vivo Evaluation of Anti-hyperlipidemic Activity.
AAPS PharmSciTech (2018-04-18)
Tejal Rawal, Neha Mishra, Abhishek Jha, Apurva Bhatt, Rajeev K Tyagi, Shital Panchal, Shital Butani
ABSTRACT
The elevated blood levels of cholesterol and low-density lipoproteins result in hyperlipidemia. The available expensive prophylactic treatments are kindred with severe side effects. Therefore, we fabricated the polymeric nanoparticles of gamma-oryzanol to achieving the improved efficacy of drug. The nanoparticles were prepared by ionic gelation method and optimized using 23 full factorial design taking drug/polymer ratio (X1), polymer/cross linking agent ratio (X2), and stirring speed (X3) as independent variables. The average particle size, percentage entrapment efficiency, and in vitro drug release at 2, 12, and 24 h were selected as response parameters. The factorial batches were statistically analyzed and optimized. The optimized nanoparticles were characterized with respect to particle size (141 nm) and zeta potential (+ 6.45 mV). Results obtained with the prepared and characterized formulation showed 83% mucoadhesion towards the intestinal mucosa. The in vitro findings were complemented well by in vivo anti-hyperlipidemic activity of developed formulation carried out in Swiss albino mouse model. The in vivo studies showed improved atherogenic index, malondialdehyde, and superoxide dismutase levels in poloxamer-407-induced hyperlipidemic animals when treated with oryzanol and gamma-oryzanol nanoformulation. Based on our findings, we believe that chitosan-mediated delivery of gamma-oryzanol nanoparticles might prove better in terms of anti-hyperlipidemic therapeutics.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)