Skip to Content
Merck
Trizma® base reference material for titrimetry, certified by BAM, >99.5%

Trizma® base

Trizma® base reference material for titrimetry, certified by BAM, >99.5%
Synonym(s):
2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Special Grade
Technique