Skip to Content
Merck
Phenoxyacetic acid PESTANAL®, analytical standard

Phenoxyacetic acid

Phenoxyacetic acid PESTANAL®, analytical standard
Synonym(s):
Glycolic acid phenyl ether
Linear Formula:
C6H5OCH2CO2H
CAS Number:
Molecular Weight:
152.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing