Skip to Content
Merck
Nitron for spectrophotometric det. of nitrate and perchlorate, ≥97.0%

Nitron

Nitron for spectrophotometric det. of nitrate and perchlorate, ≥97.0%
Synonym(s):
1,4-Diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 3,5,6-Triphenyl-2,3,5,6-tetraazabicyclo[2.1.1]hex-1-ene, 4,5-Dihydro-2,4-diphenyl-5-(phenylimino)-1H-1,2,4-triazolium hydroxide inner salt
Empirical Formula (Hill Notation):
C20H16N4
CAS Number:
Molecular Weight:
312.37
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing