Mianserin for system suitability
Synonym(s):
Mianserin hydrochloride, 1,2,3,4,10,14b-Hexahydro-2-methyl-dibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride
Empirical Formula (Hill Notation):
C18H20N2 · HCl
CAS Number:
Molecular Weight:
300.83
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing