Skip to Content
Merck
Methyl all-cis-7,10,13,16,19-docosapentaenoate ≥97%, liquid

Methyl all-cis-7,10,13,16,19-docosapentaenoate

Methyl all-cis-7,10,13,16,19-docosapentaenoate ≥97%, liquid
Synonym(s):
cis-7,10,13,16,19-Docosapentaenoic acid methyl ester
Empirical Formula (Hill Notation):
C23H36O2
CAS Number:
Molecular Weight:
344.53

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique