![Gabapentin impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/351/889/b0c0746b-fc45-4c64-b008-4ba625b440c1/640/b0c0746b-fc45-4c64-b008-4ba625b440c1.png)
Gabapentin impurity A
![Gabapentin impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/351/889/b0c0746b-fc45-4c64-b008-4ba625b440c1/640/b0c0746b-fc45-4c64-b008-4ba625b440c1.png)
Synonym(s):
4,4-Pentamethylene-2-pyrrolidinone, 3-Azaspiro[4.5]decan-2-one
Empirical Formula (Hill Notation):
C9H15NO
CAS Number:
Molecular Weight:
153.22
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing