Skip to Content
Merck
vibrant-m

Etofenamate for peak identification

vibrant-m
Synonym(s):
Etofenamate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid 2-(2-hydroxyethoxy)ethyl ester
Empirical Formula (Hill Notation):
C18H18F3NO4
CAS Number:
Molecular Weight:
369.34

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1