Skip to Content
Merck
vibrant-m

Erythromycin C

vibrant-m
Synonym(s):
3″-O-Demethylerythromycin
Empirical Formula (Hill Notation):
C36H65NO13
CAS Number:
Molecular Weight:
719.90

Products

Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade