Skip to Content
Merck
Endo-4-Methoxyphenyl Kwon [2.2.1] Bicyclic Phosphine

Endo-4-Methoxyphenyl Kwon [2.2.1] Bicyclic Phosphine

Endo-4-Methoxyphenyl Kwon [2.2.1] Bicyclic Phosphine
Synonym(s):
(1S,4S,5S)-5-(4-methoxyphenyl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C19H22NO3PS
Molecular Weight:
375.42

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing