Enanthic Acid
Synonym(s):
Heptanoic acid, Enanthic acid, Oenanthic acid
Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2