![ECO BRIJ® O10](/deepweb/assets/sigmaaldrich/product/structures/188/479/73c8b223-e813-4ae5-90f3-4634551c8c54/640/73c8b223-e813-4ae5-90f3-4634551c8c54.png)
ECO BRIJ® O10
![ECO BRIJ® O10](/deepweb/assets/sigmaaldrich/product/structures/188/479/73c8b223-e813-4ae5-90f3-4634551c8c54/640/73c8b223-e813-4ae5-90f3-4634551c8c54.png)
Synonym(s):
Brij® O10, Brij 97, C18-1E10, Polyoxyethylene (10) oleyl ether
Linear Formula:
C18H35(OCH2CH2)nOH, n~10
CAS Number:
Products
Product Category
Brand
Sustainability
Feature
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing