![(±)-2-Amino-4-phosphonobutyric acid solid](/deepweb/assets/sigmaaldrich/product/structures/395/198/7a571533-1417-46fd-a110-c5927f788054/640/7a571533-1417-46fd-a110-c5927f788054.png)
(±)-2-Amino-4-phosphonobutyric acid
![(±)-2-Amino-4-phosphonobutyric acid solid](/deepweb/assets/sigmaaldrich/product/structures/395/198/7a571533-1417-46fd-a110-c5927f788054/640/7a571533-1417-46fd-a110-c5927f788054.png)
Synonym(s):
(±)-AP-4
Empirical Formula (Hill Notation):
C4H10NO5P
CAS Number:
Molecular Weight:
183.10
Products
Brand
Color
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing