Skip to Content
Merck
Dihydroresveratrol ≥95% (HPLC)

Dihydroresveratrol

Dihydroresveratrol ≥95% (HPLC)
Synonym(s):
Dihydroresveratrol, 3,4′,5-Trihydroxybibenzyl, 3,5,4′-Trihydroxybibenzyl
Empirical Formula (Hill Notation):
C14H14O3
CAS Number:
Molecular Weight:
230.26

Products

Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing