Skip to Content
Merck
D-Cycloserine synthetic

D-Cycloserine

D-Cycloserine synthetic
Synonym(s):
(R)-4-Amino-3-isoxazolidone, 4-Amino-3-isoxazolidinone
Empirical Formula (Hill Notation):
C3H6N2O2
CAS Number:
Molecular Weight:
102.09
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Optical Isomer
Amino Acid
Showing 1-3 of 3
Compare
Product Number
Description
Pricing