Skip to Content
Merck
D-Camphor analytical standard

D-Camphor

D-Camphor analytical standard
Synonym(s):
2-Bornanone, 2-Camphanone
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique