Skip to Content
Merck
Crocin phyproof® Reference Substance

Crocin

Crocin phyproof® Reference Substance
Synonym(s):
Crocetin digentiobiose ester
Empirical Formula (Hill Notation):
C44H64O24
CAS Number:
Molecular Weight:
976.96
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing