Codergocrine mesilate
Synonym(s):
Ergoloid mesylate salt, Dihydroergotoxin mesylate salt
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing