Clonazepam impurity B
Synonym(s):
3-Amino-4-(2-chlorophenyl)-6-nitro-2(1H)-quinolinone
Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing