Chromolith® WP 300 RP-18 (monolithic) HPLC Columns
Products
Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Pore size (µm)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing