Skip to Content
Merck
Butyraldehyde-2,4-DNPH certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Butyraldehyde-2,4-DNPH

Butyraldehyde-2,4-DNPH certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Butyraldehyde-2,4-dinitrophenylhydrazone
Linear Formula:
CH3CH2CH2CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
252.23

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Reference Material Type
Special Grade