Skip to Content
Merck
Butyl alcohol ≥99.5%, FCC, FG

Butyl alcohol

Butyl alcohol ≥99.5%, FCC, FG
Synonym(s):
1-Butanol, n-Butanol, Butyl alcohol
Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing