Butralin
Synonym(s):
N-sec-Butyl-4-tert-butyl-2,6-dinitroaniline
Empirical Formula (Hill Notation):
C14H21N3O4
CAS Number:
Molecular Weight:
295.33
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing