
(−)-Bilobalide

Synonym(s):
Bilobalide
Empirical Formula (Hill Notation):
C15H18O8
CAS Number:
Molecular Weight:
326.30
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1