Skip to Content
Merck
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride ≥97.0% (HPLC)

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride ≥97.0% (HPLC)
Synonym(s):
AEBSF
Empirical Formula (Hill Notation):
C8H10FNO2S · HCl
CAS Number:
Molecular Weight:
239.69

Products

Brand
Biological Source
Feature
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing