3-Hexylthiophene
Empirical Formula (Hill Notation):
C10H16S
CAS Number:
Molecular Weight:
168.30
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing