
(2S,3R,4S)-α-(Carboxycyclopropyl)glycine

Synonym(s):
(2S,1′R,2′S)-2-(2-Carboxycyclopropyl)glycine, (2S,3R,4S)-CCG, L-CCG-IV
Empirical Formula (Hill Notation):
C6H9NO4
CAS Number:
Molecular Weight:
159.14
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing