Skip to Content
Merck
2-(2,4-Difluorophenyl)-1,2,3-propanetriol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-(2,4-Difluorophenyl)-1,2,3-propanetriol

2-(2,4-Difluorophenyl)-1,2,3-propanetriol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(2,4-Difluorophenyl)propane-1,2,3-triol
Empirical Formula (Hill Notation):
C9H10F2O3
CAS Number:
Molecular Weight:
204.17

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade