
2,2,2-Trichloro-N,N-dimethyl-13C2-acetamide

Synonym(s):
2,2,2-Trichloro-N,N-dimethyl-13C2-acetamide
Empirical Formula (Hill Notation):
13C2C2H6Cl3NO
Molecular Weight:
192.44
Products
Product Category
Formula Weight
Physical Form
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing