![1,4-Bis(2-ethylhexyl)benzene certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/251/252/9005dde6-ce61-4271-b8d2-d015c364f8e6/640/9005dde6-ce61-4271-b8d2-d015c364f8e6.png)
1,4-Bis(2-ethylhexyl)benzene
![1,4-Bis(2-ethylhexyl)benzene certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/251/252/9005dde6-ce61-4271-b8d2-d015c364f8e6/640/9005dde6-ce61-4271-b8d2-d015c364f8e6.png)
Linear Formula:
C6H4[CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
302.54
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing