Skip to Content
Merck
All Photos(1)

Key Documents

HG000C16

Millipore

Tryptone Soy Broth

(Casein-peptone Soymeal-peptone Broth), pH 7.3±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

composition

Dextrose, 2.5 g/L
di-Potassium Hydrogen Phosphate, 2.5 g/L
Sodium Chloride, 5.0 g/L
Soy Peptone, 5.0 g/L
Tryptone, 15.0 g/L

packaging

pkg of 5 kg (HG000C16.5)
poly bottle of 12 × 500 g (HG000C16.500)

pH

7.3±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A versatile liquid medium suitable for general laboratory use. Due to its high nutrient value, it will produce luxuriant growth of many fastidious organisms without the addition of serum.

Preparation Note

Suspend 30g in 1 litre demineralised water. Dispense into final containers and autoclave at 121°C for 15 minutes.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service