Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BSS-1005

Sigma-Aldrich

Dulbecco′s Phosphate Buffered Saline, 1X ES Cell Qualified

1X, liquid, ES Cell Qualifie

Synonym(s):

D-PBS, DPBS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

Product Name

Dulbecco′s Phosphate Buffered Saline, 1X ES Cell Qualified, The Dulbecco′s Phosphate Buffered Saline, 1X ES Cell Qualified is available in a 500 mL format & has been optimized & validated for Stem cell culture.

form

liquid

manufacturer/tradename

Specialty Media

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)
sample type cardiac stem cell(s)
sample type: human embryonic stem cell(s)

General description

Dulbecco′s Phosphate Buffered Saline, 1X ES Cell Qualified

Storage and Stability

Store at 15°C-30°C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Protocols

    Stem Cell protocols for cryopreservation, thawing of cryopreserved stem cells and media preparation.

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service