Skip to Content
Merck
All Photos(1)

Key Documents

JWPH95E08C94

J & W Pharmlab LLC

Oxazol-2-yl-carbamic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
184.19
MDL number:

Iupac Name

tert-butyl oxazol-2-ylcarbamate

Inchi Code

1S/C8H12N2O3/c1-8(2,3)13-7(11)10-6-9-4-5-12-6/h4-5H,1-3H3,(H,9,10,11)

InChI key

OXROIRWYICUEMZ-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Light Yellow Solid

Product Link

https://www.jwpharmlab.com/home/product/15R0694

MSDS

https://www.jwpharmlab.com/home/product/15R0694


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service