AMBH990F5EDF
Ambeed, Inc.
2-Amino-3-methoxypropanamide hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-amino-3-methoxypropanamide hydrochloride
Inchi Code
1S/C4H10N2O2.ClH/c1-8-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H2,6,7);1H
InChI key
BGRFAWSLDPCYIA-UHFFFAOYSA-N
storage temp.
Keep in dark place,Inert atmosphere,Room temperature
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/17463-68-2.html
Synonym
2-Amino-3-methoxypropanamide hydrochloride
Signal Word
Warning
Hazard Statements
MSDS
https://www.ambeed.com/products/17463-68-2.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service