Skip to Content
Merck
All Photos(1)

Key Documents

900156

Sigma-Aldrich

UV curable resin

greener alternative

for protective coatings, optical, and outdoor applications

Synonym(s):

CPS 1020 UV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

Quality Level

form

liquid (colorless)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

viscosity

470 cP (Brookfield LDTDV-II digital viscometer with size 31 spindle)

transition temp

flash point >110 °C

density

1.1 g/mL

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Storage:
  • The storage temperature should not exceed 35 °C.
  • Keep tightly closed. Store in a cool, dry, well-ventilated area.
  • Do not expose to ultraviolet light.
  • High temperatures (>80 °C), pressure and contamination with peroxides may result in auto-polymerization of the product.
  • Unstable upon depletion of inhibitor.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and thus aligns with "Design for energy efficiency". Click here for more information.

Application

  • Cures rapidly to form a glassy, high modulus, and optically clear material.
  • Excellent for use in protective coatings, optical, and outdoor applications.
  • Non-yellowing.
  • Low oxygen inhibition.
  • Mechanical properties after curing:
Modulus*: 1660MPa
Strength*: 40MPa
Elongation*: 2.5%
Tack Free**: 130 fpm


*Test resources: Tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width.
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material Data using ASTM D 638-02a.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service