Merck
  • Enhanced performance of the microalga Chlorella sorokiniana remotely induced by the plant growth-promoting bacteria Azospirillum brasilense and Bacillus pumilus.
Enhanced performance of the microalga Chlorella sorokiniana remotely induced by the plant growth-promoting bacteria Azospirillum brasilense and Bacillus pumilus.
Scientific reports (2017-02-02)
Edgar Amavizca, Yoav Bashan, Choong-Min Ryu, Mohamed A Farag, Brad M Bebout, Luz E de-Bashan
ABSTRACT
Remote effects (occurring without physical contact) of two plant growth-promoting bacteria (PGPB) Azospirillum brasilense Cd and Bacilus pumilus ES4 on growth of the green microalga Chlorella sorokiniana UTEX 2714 were studied. The two PGPB remotely enhanced the growth of the microalga, up to six-fold, and its cell volume by about three-fold. In addition to phenotypic changes, both bacteria remotely induced increases in the amounts of total lipids, total carbohydrates, and chlorophyll a in the cells of the microalga, indicating an alteration of the microalga's physiology. The two bacteria produced large amounts of volatile compounds, including CO
Product Description
Supelco
SPME Fiber Assembly 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex fiber, for use with manual holder