Merck
  • Polysialic acid-modifying liposomes for efficient delivery of epirubicin, in-vitro characterization and in-vivo evaluation.
Polysialic acid-modifying liposomes for efficient delivery of epirubicin, in-vitro characterization and in-vivo evaluation.
International journal of pharmaceutics (2016-11-05)
Ting Zhang, Songlei Zhou, Ling Hu, Bo Peng, Yang Liu, Xiang Luo, Yanzhi Song, Xinrong Liu, Yihui Deng
ABSTRACT
Polysialic acid (PSA) serves as a hydrophilic polymer and affords conjugated biologically active molecules a longer circulation time in vivo. Furthermore, PSA could potentially target tumor tissues and help achieve better curative effects. In this study, PSA was conjugated with octadecyl dimethyl betaine (BS18) to yield a PSA-BS18 conjugate. The PSA-BS18 modified liposomal epirubicin (EPI-SL), had a particle size of 133.63±0.92nm, a zeta potential of -26.23±1.50mV and an encapsulation efficiency (%EE) of 96.23±1.16%. In vitro release studies showed that PSA-BS18 could delay EPI release from the modified liposomes. The MTT assay suggested that EPI-SL led to stronger cytotoxic activity than that exhibited by common and PEGylated liposomes. The pharmacokinetic study showed that EPI-SL prolonged the residence time of the EPI in the blood compared with that observed from common liposomes. Bio-distribution results obtained from tumor-bearing mice clearly demonstrated that PSA-BS18 increased the accumulation of modified liposomes in tumors compared with that of common liposomes. In the antitumor efficacy study, EPI-SL showed the best antitumor and life-prolonging effects among all of the tested formulations. These findings strongly indicate EPI-SL might have great potential as an effective approach for anticancer therapy.
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Pricing and availability is not currently available.