Merck
  • Dietary restriction during development enlarges intestinal and hypodermal lipid droplets in Caenorhabditis elegans.
Dietary restriction during development enlarges intestinal and hypodermal lipid droplets in Caenorhabditis elegans.
PloS one (2012-11-28)
Daniela Palgunow, Maja Klapper, Frank Döring
ABSTRACT
Dietary restriction (DR) extends lifespan in man species and modulates evolutionary conserved signalling and metabolic pathways. Most of these studies were done in adult animals. Here we investigated fat phenotypes of C. elegans larvae and adults which were exposed to DR during development. This approach was named "developmental-DR" (dDR). Moderate as well as stringent dDR increased the triglyceride to protein ratio in L4 larvae and adult worms. This alteration was accompanied by a marked expansion of intestinal and hypodermal lipid droplets. In comparison to ad libitum condition, the relative proportion of fat stored in large lipid droplets (>50 µm(3)) was increased by a factor of about 5 to 6 in larvae exposed to dDR. Microarray-based expression profiling identified several dDR-regulated genes of lipolysis and lipogenesis which may contribute to the observed fat phenotypes. In conclusion, dDR increases the triglyceride to protein ratio, enlarges lipid droplets and alters the expression of genes functioning in lipid metabolism in C. elegans. These changes might be an effective adaptation to conserve fat stores in animals subjected to limiting food supply during development.
Product Description
Supelco
Phospholipid mixture for HPLC from Glycine max (soybean), certified reference material
Pricing and availability is not currently available.