Merck
  • A solid-phase extraction method for rapidly determining the adsorption coefficient of pharmaceuticals in sewage sludge.
A solid-phase extraction method for rapidly determining the adsorption coefficient of pharmaceuticals in sewage sludge.
Water research (2014-10-10)
Laurence Berthod, Gary Roberts, David C Whitley, Alan Sharpe, Graham A Mills
ABSTRACT
The partitioning of pharmaceuticals in the environment can be assessed by measuring their adsorption coefficients (Kd) between aqueous and solid phases. Measuring this coefficient in sewage sludge gives an indication of their partitioning behaviour in a wastewater treatment plant and hence contributes to an understanding of their subsequent fate. The regulatory approved method for measuring Kd in sewage sludge is the US Environmental Protection Agency's Office of Prevention, Pesticides and Toxic Substances (OPPTS) guideline 835.1110, which is labour intensive and time consuming. We describe an alternative method for measuring the Kd of pharmaceuticals in sewage sludge using a modified solid-phase extraction (SPE) technique. SPE cartridges were packed at different sludge/PTFE ratios (0.4, 6.0, 24.0 and 40.0% w/w sludge) and eluted with phosphate buffer at pH 7.4. The approach was tested initially using three pharmaceuticals (clofibric acid, diclofenac and oxytetracycline) that covered a range of Kd values. Subsequently, the sorption behaviour of ten further pharmaceuticals with varying physico-chemical properties was evaluated. Results from the SPE method were comparable to those of the OPPTS test, with a correlation coefficient of 0.93 between the two approaches. SPE cartridges packed with sludge and PTFE were stable for up to one year; use within one month reduced variability in measurements (to a maximum of 0.6 log units). The SPE method is low-cost, easy to use and enables the rapid measurement of Kd values for a large number of chemicals. It can be used as an alternative to the more laborious full OPPTS test in environmental fate studies and risk assessments.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium phosphate, 96%
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)