Merck
  • One-pot enzymatic production of beta-D-galactopyranosyl-(1-->3)-2-acetamido-2-deoxy-D-galactose (galacto-N-biose) from sucrose and 2-acetamido-2-deoxy-D-galactose (N-acetylgalactosamine).
One-pot enzymatic production of beta-D-galactopyranosyl-(1-->3)-2-acetamido-2-deoxy-D-galactose (galacto-N-biose) from sucrose and 2-acetamido-2-deoxy-D-galactose (N-acetylgalactosamine).
Carbohydrate research (2009-10-24)
Mamoru Nishimoto, Motomitsu Kitaoka
ABSTRACT
Beta-D-Galactopyranosyl-(1-->3)-2-acetamido-2-deoxy-D-galactose (galacto-N-biose, GNB) is an important core structure in functional sugar chains such as T-antigen disaccharide and the core 1 sugar chain in mucin glycoproteins. We successfully developed a one-pot enzymatic production of GNB from sucrose and GalNAc by the concomitant action of four enzymes: sucrose phosphorylase, UDP-glucose-hexose 1-phosphate uridylyltransferase, UDP-glucose 4-epimerase, and galacto-N-biose/lacto-N-biose I phosphorylase in the presence of UDP-glucose and phosphate, by modifying the method of lacto-N-biose I production [Nishimoto, M.; Kitaoka, M., Biosci. Biotechnol. Biochem., 2007, 71, 2101-2104]. The reaction yield of GNB was 88% from GalNAc. GNB was isolated from the reaction mixture by crystallization after yeast treatment to obtain approximately 45 g of GNB in 95% purity from a 280-mL reaction mixture.
Sigma-Aldrich
Sucrose Phosphorylase, recombinant, expressed in E. coli, lyophilized powder, ≥45 units/mg solid