Merck
  • Regeneration of pulp-dentin complex using human stem cells of the apical papilla: in vivo interaction with two bioactive materials.
Regeneration of pulp-dentin complex using human stem cells of the apical papilla: in vivo interaction with two bioactive materials.
Clinical oral investigations (2021-02-26)
Diana B Sequeira, Ana Rafaela Oliveira, Catarina M Seabra, Paulo J Palma, Carlos Ramos, Maria H Figueiredo, Ana C Santos, Ana L Cardoso, João Peça, João Miguel Santos
ABSTRACT
To compare the regenerative properties of human stem cells of the apical papilla (SCAPs) embedded in a platelet-rich plasma (PRP) scaffold, when implanted in vivo using an organotypic model composed of human root segments, with or without the presence of the bioactive cements - ProRoot MTA or Biodentine. SCAPs were isolated from third molars with incomplete rhizogenesis and expanded and characterized in vitro using stem cell and surface markers. The pluripotency of these cells was also assessed using adipogenic, chondrogenic, and osteogenic differentiation protocols. SCAPs together with a scaffold of PRP were added to the root segment lumen and the organotypic model implanted on the dorsal region of immunodeficient rats for a period of 4 months. Presence of SCAPs induced de novo formation of dentin-like and pulp-like tissue. A barrier of either ProRoot MTA or Biodentine did not significantly affect the fraction of sections from roots segments observed to contain deposition of hard material (P > 0.05). However, the area of newly deposited dentin was significantly greater in segments containing a barrier of Biodentine compared with ProRoot MTA (P < 0.001). SCAPs offer a viable alternative to other dental stem cells (DSCs) in their regenerative properties when enclosed in the microenvironment of human tooth roots. The present study also suggests that the presence of bioactive materials does not hinder or impede the formation of new hard tissues, but the presence of Biodentine may promote greater mineralized tissue deposition.
Sigma-Aldrich
Paraplast®, for tissue embedding