Merck
  • Time-Dependent Changes in Hepatic Sphingolipid Accumulation and PI3K/Akt/mTOR Signaling Pathway in a Rat Model of NAFLD.
Time-Dependent Changes in Hepatic Sphingolipid Accumulation and PI3K/Akt/mTOR Signaling Pathway in a Rat Model of NAFLD.
International journal of molecular sciences (2021-11-28)
Klaudia Sztolsztener, Karolina Konstantynowicz-Nowicka, Ewa Harasim-Symbor, Adrian Chabowski
ABSTRACT
Increased lipid bioavailability in a diet favors lipid accumulation, enhancing hepatic lipotoxicity and contributing to insulin resistance (IR) development. The aim of our study was to examine time-dependent alterations in the intrahepatic content of sphingolipids and insulin signaling pathway in rats fed a high-fat diet (HFD). The experiment was conducted on male Wistar rats receiving a standard diet or HFD for five weeks. At the end of each experimental feeding week, liver sphingolipids were determined using high-performance liquid chromatography. The expression of proteins from the sphingolipid pathway and glucose transporter expression were assessed by Western blot. The content of phosphorylated form of proteins from the insulin pathway was detected by a multiplex assay kit. Our results revealed that HFD enhanced hepatic ceramide deposition by increasing the expression of selected proteins from sphingomyelin and salvage pathways in the last two weeks. Importantly, we observed a significant inhibition of Akt phosphorylation in the first week of HFD and stimulation of PTEN and mTOR phosphorylation at the end of HFD. These changes worsened the PI3K/Akt/mTOR signaling pathway. We may postulate that HFD-induced reduction in the insulin action in the time-dependent matter was exerted by excessive accumulation of sphingosine and sphinganine rather than ceramide.
Sigma-Aldrich
Anti-SPHK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SPHK2 antibody produced in rabbit, affinity isolated antibody